Assay Details
Target Gene Details
Entrez Gene ID: | 269854 |
Gene Name: | N-acetyltransferase 14 |
Gene Aliases: |
BC047219 |
Location: |
Chr.7:4918888-4925006 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nat14 | NM_201355.3 | 1 | 312 | NP_958743.1 |
| AK078272.1 | BAC37199.1 | |||
| AK158627.1 | 1 | 313 | BAE34587.1 |
More Information
Set Membership: |
|

Genomic Map