Assay Details
Target Gene Details
Entrez Gene ID: | 12606 |
Gene Name: | CCAAT/enhancer binding protein (C/EBP), alpha |
Gene Aliases: |
C/ebpalpha, CBF-A, Cebp |
Location: |
Chr.7:35119293-35121928 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cebpa | NM_001287514.1 | 1 | 2197 | NP_001274443.1 |
| NM_001287515.1 | 1 | 2197 | NP_001274444.1 | |
| NM_001287521.1 | 1 | 2197 | NP_001274450.1 | |
| NM_001287523.1 | 1 | 2197 | NP_001274452.1 | |
| NM_007678.3 | 1 | 2197 | NP_031704.2 | |
| AK155281.1 | 1 | 2157 | BAE33162.1 | |
| BC011118.1 | 1 | 2187 | AAH11118.1 | |
| BC028890.1 | AAH28890.1 | |||
| BC051102.1 | 1 | 2187 | AAH51102.1 | |
| BC058161.1 | 1 | 2187 | AAH58161.1 |
More Information
Set Membership: |
|

Genomic Map