Assay Details
Target Gene Details
Entrez Gene ID: | 69358 |
Gene Name: | leucine rich repeat containing 51 |
Gene Aliases: |
1700008D07Rik, Lrtomt |
Location: |
Chr.7:101912989-101933917 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrrc51 | NM_001162973.1 | NP_001156445.1 | ||
NM_001162974.1 | 3 | 647 | NP_001156446.1 | |
NM_027053.1 | NP_081329.1 | |||
AK005758.1 | BAB24223.1 | |||
BC025128.1 | 3 | 698 | AAH25128.1 | |
EU627089.1 |
