Assay Details
Target Gene Details
Entrez Gene ID: | 213573 |
Gene Name: | calcium release activated channel regulator 2B |
Gene Aliases: |
6330520A15Rik, BC026645, Cracr2a, Efcab4, Efcab4a |
Location: |
Chr.7:141460874-141466613 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cracr2b | NM_001025103.2 | 10 | 1407 | NP_001020274.2 |
| BC026645.1 | 7 | 981 | AAH26645.1 | |
| BC034124.1 | 4 | 603 | AAH34124.1 | |
| BC048916.1 | 10 | 1407 | AAH48916.1 | |
| BC098333.1 | 10 | 1368 | AAH98333.1 |

Genomic Map