Assay Details
Target Gene Details
Entrez Gene ID: | 243937 |
Gene Name: | zinc finger protein 536 |
Gene Aliases: |
9630010P11Rik, Znf536, mKIAA0390 |
Location: |
Chr.7:37318023-37774177 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp536 | AK039356.1 | |||
BC033594.1 | AAH33594.1 |
