Assay Details
Target Gene Details
Entrez Gene ID: | 83485 |
Gene Name: | neugrin, neurite outgrowth associated |
Gene Aliases: |
AW552001 |
Location: |
Chr.7:80261215-80265378 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ngrn | NM_031375.4 | 3 | 648 | NP_113552.3 |
NR_028053.1 | 2 | 1045 | ||
AB047544.1 | 3 | 674 | BAB21528.1 | |
AK083330.1 | 2 | 1041 | BAC38868.1 | |
AK144756.1 | 3 | 644 | BAE26050.1 | |
AK146059.1 | 2 | 1023 | BAE26868.1 | |
AK181653.1 | 1 | 330 |
