Assay Details
Target Gene Details
Entrez Gene ID: | 66085 |
Gene Name: | eukaryotic translation initiation factor 3, subunit F |
Gene Aliases: |
0610037M02Rik, AA409853, Eif3s5 |
Location: |
Chr.7:108934415-108941942 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3f | NM_025344.2 | 8 | 1341 | NP_079620.2 |
More Information
Set Membership: |
|

Genomic Map