Assay Details
Target Gene Details
Entrez Gene ID: | 245126 |
Gene Name: | T cell-interacting, activating receptor on myeloid cells 1 |
Gene Aliases: |
9930022N03Rik, ENSMUSG00000053338, Gm9904, OLT-2 |
Location: |
Chr.7:3486499-3502643 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tarm1 | NM_177363.3 | 4 | 553 | NP_796337.1 |
| AK036904.1 | 4 | 554 | BAC29632.1 | |
| AK155804.1 | 4 | 561 | BAE33442.1 | |
| AK170277.1 | 3 | 484 | BAE41681.1 | |
| DQ973493.1 | 4 | 505 |
More Information
Set Membership: |
|

Genomic Map