Assay Details
Target Gene Details
Entrez Gene ID: | 76932 |
Gene Name: | ADP-ribosylation factor interacting protein 2 |
Gene Aliases: |
2310002N04Rik, POR, Por1 |
Location: |
Chr.7:105634201-105640416 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arfip2 | NM_029802.4 | 8 | 2551 | NP_084078.3 |
More Information
Set Membership: |
|

Genomic Map