Assay Details
Target Gene Details
Entrez Gene ID: | 320210 |
Gene Name: | RIKEN cDNA A230077H06 gene |
Gene Aliases: |
- |
Location: |
Chr.7:40849153-40898324 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
A230077H06Rik | NR_040329.1 | |||
AK081136.1 |
Target Gene Details
Entrez Gene ID: | 58188 |
Gene Name: | V-set and transmembrane domain containing 2B |
Gene Aliases: |
2900093B09Rik, AB030198 |
Location: |
Chr.7:40898297-40930044 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vstm2b |
