Assay Details
Target Gene Details
Entrez Gene ID: | 17293 |
Gene Name: | mesoderm posterior 2 |
Gene Aliases: |
bHLHc6 |
Location: |
Chr.7:79810727-79813431 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mesp2 | NM_008589.2 | 1 | 687 | NP_032615.2 |
AK146740.1 | 1 | 167 | BAE27401.1 | |
BC145652.1 | 1 | 517 | ||
BC145654.1 | 1 | 517 | ||
U71125.1 | 1 | 693 | AAB51199.1 |
