Assay Details
Target Gene Details
Entrez Gene ID: | 101613 |
Gene Name: | NLR family, pyrin domain containing 6 |
Gene Aliases: |
AI504961, Avr, Nalp6, Navr, Navr/Avr, Pypaf5 |
Location: |
Chr.7:140920902-140929192 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nlrp6 | NM_133946.2 | 4 | 1074 | NP_598707.2 |
AK039531.1 | 1 | 358 | ||
AY355343.1 | AAR14740.1 | |||
BC013519.1 | AAH13519.1 | |||
BC031139.1 | 1 | 66 | AAH31139.1 | |
FJ222825.1 | 4 | 1023 | ||
JF810536.1 | 3 | 1459 |
