Assay Details
Target Gene Details
Entrez Gene ID: | 101613 |
Gene Name: | NLR family, pyrin domain containing 6 |
Gene Aliases: |
AI504961, Avr, Nalp6, Navr, Navr/Avr, Pypaf5 |
Location: |
Chr.7:140920902-140929192 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nlrp6 | NM_133946.2 | 8 | 2855 | NP_598707.2 |
AK039531.1 | 5 | 2139 | ||
AY355343.1 | 5 | 1793 | AAR14740.1 | |
BC013519.1 | 5 | 1762 | AAH13519.1 | |
BC031139.1 | 5 | 1847 | AAH31139.1 | |
FJ222825.1 | 8 | 2801 |
