Assay Details
Target Gene Details
Entrez Gene ID: | 211548 |
Gene Name: | nodal modulator 1 |
Gene Aliases: |
D7Ertd156e, Nomo, pM5 |
Location: |
Chr.7:46033696-46084212 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nomo1 | NM_153057.4 | 1 | 229 | NP_694697.3 |
AK030584.1 | 1 | 196 | ||
AK035957.1 | 1 | 197 | ||
AK080754.1 | 1 | 88 | ||
AK082495.1 | 1 | 108 | BAC38508.1 | |
AK082963.1 | 1 | 88 | BAC38713.1 | |
AK142338.1 | 1 | 197 | ||
AK150107.1 | 1 | 183 | ||
AK165643.1 | 1 | 90 | ||
AK192010.1 | ||||
AK195816.1 | ||||
AK220126.1 | ||||
BC072630.1 | 1 | 175 | AAH72630.1 |
