Assay Details
Target Gene Details
Entrez Gene ID: | 434198 |
Gene Name: | RIKEN cDNA B130024G19 gene |
Gene Aliases: |
- |
Location: |
Chr.7:70365383-70411146 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B130024G19Rik | NR_045850.1 | |||
| AK045070.1 | BAC32208.1 | |||
| AK051651.1 | 1 | 1622 |
Target Gene Details
Entrez Gene ID: | 11819 |
Gene Name: | nuclear receptor subfamily 2, group F, member 2 |
Gene Aliases: |
2700033K02Rik, 9430015G03Rik, ARP-1, Aporp1, COUP-TF2, COUP-TFII, COUPTFB, EAR3, SVP40, Tcfcoup2 |
Location: |
Chr.7:70351946-70366746 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr2f2 | NM_183261.3 | 1 | 217 | NP_899084.2 |
| BY001758.1 | 1 | 220 |
More Information
Set Membership: |
|

Genomic Map