Assay Details
Target Gene Details
Entrez Gene ID: | 66411 |
Gene Name: | tubulin folding cofactor B |
Gene Aliases: |
2410007D12Rik, AU041393, CG22, CKAPI, Ckap1 |
Location: |
Chr.7:30224130-30232029 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbcb | NM_025548.3 | 1 | 221 | NP_079824.2 |
| BQ445322.1 | 1 | 221 |
More Information
Set Membership: |
|

Genomic Map