Assay Details
Target Gene Details
Entrez Gene ID: | 259277 |
Gene Name: | kallikrein related-peptidase 8 |
Gene Aliases: |
BSP1, Nrpn, Prss19 |
Location: |
Chr.7:43797552-43803822 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klk8 | NM_008940.2 | 1 | 455 | NP_032966.1 |
| BC055895.1 | AAH55895.1 | |||
| D30785.1 | 1 | 455 | BAA06451.1 |
More Information
Set Membership: |
|

Genomic Map