Assay Details
Target Gene Details
Entrez Gene ID: | 50994 |
Gene Name: | metastasis associated gene 2 |
Gene Aliases: |
MAG |
Location: |
Chr.7:45366163-45367948 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtag2 | NR_015480.1 | |||
| AK042966.1 |
Target Gene Details
Entrez Gene ID: | 76787 |
Gene Name: | protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 3 |
Gene Aliases: |
2410127E16Rik |
Location: |
Chr.7:45339126-45367019 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppfia3 | NM_029741.2 | 1 | 124 | NP_084017.2 |
| AK147645.1 | 1 | 124 | ||
| BC058404.1 | AAH58404.1 | |||
| EU568871.1 |
More Information
Set Membership: |
|

Genomic Map