Assay Details
Target Gene Details
Entrez Gene ID: | 69714 |
Gene Name: | TCF3 (E2A) fusion partner |
Gene Aliases: |
2400004F01Rik, AI450389, Amida, FB1 |
Location: |
Chr.7:3620324-3629929 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tfpt | NM_001290381.1 | 2 | 508 | NP_001277310.1 |
NM_023524.2 | 2 | 508 | NP_076013.1 | |
AB035134.1 | 2 | 199 | BAB17318.1 | |
AF267988.1 | 2 | 463 | AAG48243.1 | |
AK010279.1 | 2 | 166 | BAB26816.1 | |
AK155925.1 | 2 | 492 | BAE33506.1 | |
BC024356.1 | 2 | 407 | AAH24356.1 | |
CX768547.1 |
