Assay Details
Target Gene Details
Entrez Gene ID: | 233231 |
Gene Name: | MAS-related GPR, member B1 |
Gene Aliases: |
MrgB1 |
Location: |
Chr.7:48444113-48456342 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgprb1 | NM_205810.4 | 2 | 3516 | NP_991379.3 |
| AK148046.1 | 2 | 3421 | ||
| AK148074.1 | 2 | 3517 | BAE28326.1 | |
| AK148108.1 | 2 | 3421 | BAE28348.1 | |
| AK148256.1 | BAE28442.1 | |||
| AK148279.1 | 2 | 3470 | BAE28455.1 |
More Information
Set Membership: |
|

Genomic Map