Assay Details
Target Gene Details
Entrez Gene ID: | 435965 |
Gene Name: | low density lipoprotein receptor-related protein 3 |
Gene Aliases: |
- |
Location: |
Chr.7:35200878-35215493 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrp3 | NM_001024707.2 | 8 | 3829 | NP_001019878.1 |
| AK149314.1 | 7 | 3354 | BAE28807.1 | |
| BC094349.1 | 2 | 1923 | AAH94349.1 | |
| CJ194460.1 | 1 | 230 |
Target Gene Details
Entrez Gene ID: | 53896 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 10 |
Gene Aliases: |
AL024237, Asc-1, D7Bwg0847e |
Location: |
Chr.7:35186357-35201114 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc7a10 | NM_017394.4 | 11 | 1882 | NP_059090.3 |
| AK031867.1 | 11 | 1850 | BAC27582.1 | |
| BC054765.1 | 11 | 1825 | AAH54765.1 |
More Information
Set Membership: |
|

Genomic Map