Assay Details
Target Gene Details
Entrez Gene ID: | 71829 |
Gene Name: | DDI1, DNA-damage inducible 1, homolog 1 (S. cerevisiae) |
Gene Aliases: |
1700011N24Rik |
Location: |
Chr.9:6265028-6266547 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddi1 | NM_027942.1 | 1 | 514 | NP_082218.1 |
| AK005882.1 | 1 | 516 | BAB24297.1 |
Target Gene Details
Entrez Gene ID: | 71785 |
Gene Name: | platelet-derived growth factor, D polypeptide |
Gene Aliases: |
1110003I09Rik |
Location: |
Chr.9:6168362-6536014 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdgfd | NM_027924.2 | NP_082200.1 | ||
| AF335583.1 | AAK38839.1 | |||
| AK003359.2 | BAB22735.2 | |||
| AK141551.1 | BAE24732.1 | |||
| BC030896.1 | AAH30896.1 |
More Information
Set Membership: |
|

Genomic Map