Assay Details
Target Gene Details
Entrez Gene ID: | 102060 |
Gene Name: | growth arrest and DNA-damage-inducible, gamma interacting protein 1 |
Gene Aliases: |
2310040G17Rik, AI425883, Crif1, MRP-L59, Plinp1 |
Location: |
Chr.8:84832282-84835482 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gadd45gip1 | NM_183358.4 | 1 | 294 | NP_899202.3 |
| AK002738.1 | 1 | 300 | BAB22318.1 | |
| AK003314.1 | 1 | 295 | BAB22710.1 | |
| AK009723.1 | 1 | 293 | BAB26464.1 | |
| AK019570.1 | 1 | 295 | BAB31797.1 | |
| AK028227.1 | 1 | 295 | BAC25826.1 | |
| BC061069.1 | 1 | 294 | AAH61069.1 |

Genomic Map