Assay Details
Target Gene Details
Entrez Gene ID: | 270152 |
Gene Name: | adhesion molecule, interacts with CXADR antigen 1 |
Gene Aliases: |
AMICA, Crea7, Gm638 |
Location: |
Chr.9:45079162-45108531 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amica1 | NM_001005421.4 | 11 | 1503 | NP_001005421.3 |
| AY093688.1 | 11 | 1503 | AAM15732.1 | |
| CJ231590.1 | 1 | 188 |
Target Gene Details
Entrez Gene ID: | 100038468 |
Gene Name: | predicted gene 10684 |
Gene Aliases: |
ENSMUSG00000074385 |
Location: |
Chr.9:45107176-45135606 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10684 | NR_033547.1 | 3 | 1803 | |
| AK045437.1 | 3 | 1804 | BAE20650.1 |
More Information
Set Membership: |
|

Genomic Map