Assay Details
Target Gene Details
Entrez Gene ID: | 330817 |
Gene Name: | deoxyhypusine synthase |
Gene Aliases: |
Dhs |
Location: |
Chr.8:85069718-85075162 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dhps | NM_001039514.1 | NP_001034603.1 | ||
AK159104.1 | BAE34820.1 | |||
AK183443.1 | ||||
AK212076.1 | ||||
BB763715.1 | ||||
BC039963.1 | AAH39963.1 |
Target Gene Details
Entrez Gene ID: | 67836 |
Gene Name: | WD repeat domain containing 83 |
Gene Aliases: |
1500041N16Rik, Morg1 |
Location: |
Chr.8:85075035-85080746 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wdr83 | NM_026399.2 | 9 | 1081 | NP_080675.2 |
AK005790.1 | 9 | 1021 | BAB24241.1 | |
AK020654.1 | 9 | 1042 | BAB32164.1 | |
AY365434.1 | 9 | 1081 | AAQ64635.1 | |
BC005587.1 | 7 | 1404 | AAH05587.1 | |
BC019369.1 | 9 | 1004 | AAH19369.1 | |
BC094624.1 | AAH94624.1 |
