Assay Details
Target Gene Details
Entrez Gene ID: | 20516 |
Gene Name: | solute carrier family 20, member 2 |
Gene Aliases: |
MolPit2, Pit-2, Pit2, Ram-1, Ram1 |
Location: |
Chr.8:22476700-22569616 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc20a2 | NM_011394.3 | 11 | 3420 | NP_035524.2 |
| AK087507.1 | 11 | 3245 | ||
| AK182280.1 | 1 | 195 | ||
| AK196312.1 | 1 | 120 | ||
| AK203538.1 | 1 | 120 | ||
| BC046510.1 | 11 | 3343 | AAH46510.1 |
More Information
Set Membership: |
|

Genomic Map