Assay Details
Target Gene Details
Entrez Gene ID: | 70345 |
Gene Name: | RIKEN cDNA 0610038B21 gene |
Gene Aliases: |
- |
Location: |
Chr.8:77517056-77518578 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 0610038B21Rik | NR_028125.1 | 1 | 609 | |
| AK002788.1 | 1 | 610 | BAB22359.1 |
Target Gene Details
Entrez Gene ID: | 78514 |
Gene Name: | Rho GTPase activating protein 10 |
Gene Aliases: |
A930033B01Rik, PSGAP-m, PSGAP-s |
Location: |
Chr.8:77250364-77517953 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap10 | NM_030113.2 | 1 | 244 | NP_084389.2 |
| AF297030.1 | 1 | 207 | AAK18175.1 | |
| AK020920.1 | 1 | 245 | BAB32254.3 | |
| AK041867.1 | 1 | 291 | BAC31086.1 | |
| AY179965.1 | 1 | 120 | AAO62072.1 | |
| AY179966.1 | 1 | 120 | AAO62073.1 | |
| AY179967.1 | 1 | 120 | AAO62074.1 |
More Information
Set Membership: |
|

Genomic Map