Assay Details
Target Gene Details
Entrez Gene ID: | 56773 |
Gene Name: | carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 5 |
Gene Aliases: |
AI173964, GST-4, Gn6st-3, I-GlcNAc-6-ST |
Location: |
Chr.8:111889135-111910199 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chst5 | NM_019950.2 | 2 | 1620 | NP_064334.1 |
| AF176840.1 | 2 | 1643 | AAD56002.1 |
More Information
Set Membership: |
|

Genomic Map