Assay Details
Target Gene Details
Entrez Gene ID: | 109145 |
Gene Name: | GINS complex subunit 4 (Sld5 homolog) |
Gene Aliases: |
2810037C03Rik, 4933405K01Rik, Sld5 |
Location: |
Chr.8:23226610-23237668 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gins4 | NM_024240.6 | 8 | 992 | NP_077202.1 |
| AK016667.1 | 7 | 967 | ||
| AK076104.1 | 8 | 984 | BAC36185.1 | |
| AK164815.1 | 1 | 190 | BAE37931.1 | |
| AK177256.1 | ||||
| AK211204.1 | ||||
| AK219168.1 | ||||
| BB750165.1 | 1 | 70 | ||
| BC002156.1 | 8 | 963 | AAH02156.1 |

Genomic Map