Assay Details
Target Gene Details
Entrez Gene ID: | 270096 |
Gene Name: | MON1 homolog b (yeast) |
Gene Aliases: |
5031407H10, 5033413H12Rik |
Location: |
Chr.8:113635586-113645192 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mon1b | NM_001048143.2 | 4 | 1030 | NP_001041608.1 |
| NM_173015.3 | 3 | 1220 | NP_766603.1 | |
| AK030288.1 | 3 | 1186 | BAC26880.1 | |
| AK157390.1 | 4 | 1030 | ||
| AK173047.1 | 3 | 892 | BAD32325.1 | |
| BC062639.1 | 3 | 896 | AAH62639.1 |

Genomic Map