Assay Details
Target Gene Details
Entrez Gene ID: | 11814 |
Gene Name: | apolipoprotein C-III |
Gene Aliases: |
apo-CIII, apoC-III |
Location: |
Chr.9:46233050-46235636 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apoc3 | NM_001289755.1 | 4 | 398 | NP_001276684.1 |
| NM_001289756.1 | 4 | 368 | NP_001276685.1 | |
| NM_001289833.1 | 3 | 226 | NP_001276762.1 | |
| NM_023114.4 | 4 | 294 | NP_075603.1 | |
| AK002908.1 | 4 | 293 | BAB22448.1 | |
| AK008260.1 | 4 | 297 | BAB25563.1 | |
| BC021776.1 | 4 | 276 | AAH21776.1 | |
| L04150.1 | 4 | 293 | ||
| W61891.1 | 4 | 368 |
More Information
Set Membership: |
|

Genomic Map