Assay Details
Target Gene Details
Entrez Gene ID: | 330723 |
Gene Name: | HtrA serine peptidase 4 |
Gene Aliases: |
B430206E18Rik |
Location: |
Chr.8:25024928-25038962 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Htra4 | NM_001081187.3 | 1 | 94 | NP_001074656.1 |
| AK046614.1 | ||||
| BB619816.1 | 1 | 94 | ||
| BC132380.1 | 1 | 54 | ||
| BC145842.1 | 1 | 54 |
More Information
Set Membership: |
|

Genomic Map