Assay Details
Target Gene Details
Entrez Gene ID: | 57312 |
Gene Name: | mitochondrial ribosomal protein S31 |
Gene Aliases: |
1500002D03Rik, AA960481, Imogen38, Imogen44 |
Location: |
Chr.8:22410472-22431044 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps31 | NM_020560.2 | 7 | 1128 | NP_065585.1 |
| AK005110.1 | 7 | 1087 | BAB23825.1 | |
| AK011825.1 | 7 | 1084 | ||
| AK019202.1 | 7 | 1109 | ||
| AK161913.1 | 7 | 1071 | BAE36630.1 | |
| BC018475.1 | 7 | 1027 | AAH18475.1 | |
| BC027430.1 | 7 | 1032 | AAH27430.1 | |
| Z46966.1 | 7 | 1047 | CAA87087.1 |
More Information
Set Membership: |
|

Genomic Map