Assay Details
Target Gene Details
Entrez Gene ID: | 23808 |
Gene Name: | ash2 (absent, small, or homeotic)-like (Drosophila) |
Gene Aliases: |
- |
Location: |
Chr.8:25816000-25847694 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ash2l | NM_001080793.2 | 16 | 2235 | NP_001074262.1 |
| NM_001286207.1 | 16 | 2208 | NP_001273136.1 | |
| NM_011791.3 | 16 | 2325 | NP_035921.2 | |
| AB020983.2 | 16 | 2307 | BAA35128.1 | |
| AK087934.1 | 16 | 2300 | BAC40047.1 | |
| AK132471.1 | 13 | 3208 | ||
| AK145576.1 | 16 | 2200 | BAE26521.1 | |
| AK145967.1 | 16 | 2179 | BAE26792.1 | |
| AK146254.1 | 16 | 2237 | BAE27016.1 | |
| AK146599.1 | 16 | 2209 | BAE27293.1 | |
| AK146756.1 | 16 | 2200 | BAE27411.1 | |
| AK146938.1 | 16 | 2302 | BAE27547.1 | |
| AK213310.1 | 1 | 228 | ||
| BC012957.1 | 16 | 2312 | AAH12957.1 | |
| BE136411.1 | 1 | 111 | ||
| CV556779.1 | 1 | 139 |
More Information
Set Membership: |
|

Genomic Map