Assay Details
Target Gene Details
Entrez Gene ID: | 234776 |
Gene Name: | ATM interactor |
Gene Aliases: |
Asciz, gpg6, mKIAA0431 |
Location: |
Chr.8:116943386-116960445 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atmin | NM_177700.4 | 4 | 1384 | NP_808368.3 |
AK087334.1 | 1 | 695 | BAC39849.1 | |
AK122281.1 | 4 | 1266 | BAC65563.1 | |
AX775769.1 | 4 | 1384 | CAE11584.1 | |
BC060631.1 | 4 | 1369 | AAH60631.1 |
