Assay Details
Target Gene Details
Entrez Gene ID: | 192156 |
Gene Name: | mevalonate (diphospho) decarboxylase |
Gene Aliases: |
C78718, Mpd |
Location: |
Chr.8:122433596-122443422 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mvd | NM_138656.2 | 10 | 1656 | NP_619597.2 |
| NR_028354.1 | 11 | 1759 | ||
| AJ309922.1 | 10 | 1613 | CAC35731.1 | |
| AK089354.1 | 10 | 1658 | BAC40852.1 | |
| AK134803.1 | 10 | 1645 | BAE22291.1 | |
| AK153439.1 | 11 | 1734 | ||
| AK154883.1 | 10 | 1658 | BAE32901.1 | |
| AK187633.1 | 1 | 65 | ||
| AK212634.1 | 1 | 65 | ||
| BC008526.1 | 10 | 1624 | AAH08526.1 |
More Information
Set Membership: |
|

Genomic Map