Assay Details
Target Gene Details
Entrez Gene ID: | 66595 |
Gene Name: | asteroid homolog 1 (Drosophila) |
Gene Aliases: |
1100001A21Rik, AV006403 |
Location: |
Chr.9:105395294-105408298 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aste1 | NM_025651.4 | 2 | 1366 | NP_079927.2 |
| AK003146.1 | 1 | 716 | BAB22602.1 | |
| AK030661.1 | 2 | 1237 | BAC27067.1 | |
| BC098471.1 | 3 | 1251 | AAH98471.1 |

Genomic Map