Assay Details
Target Gene Details
Entrez Gene ID: | 20845 |
Gene Name: | steroidogenic acute regulatory protein |
Gene Aliases: |
AV363654, D8Ertd419e |
Location: |
Chr.8:25808513-25815982 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Star | NM_011485.4 | 7 | 1331 | NP_035615.2 |
AK014431.1 | 7 | 1340 | ||
AK019725.2 | 7 | 1371 | BAB31842.1 | |
AK054470.1 | 7 | 1331 | BAC35791.1 | |
BC060970.1 | AAH60970.1 | |||
BC082283.1 | 7 | 1342 | AAH82283.1 | |
L36062.1 | 7 | 1347 | AAB94783.1 |
