Assay Details
Target Gene Details
Entrez Gene ID: | 67384 |
Gene Name: | BCL2-associated athanogene 4 |
Gene Aliases: |
2410112I15Rik, SODD |
Location: |
Chr.8:25764538-25785209 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bag4 | NM_026121.3 | 4 | 805 | NP_080397.1 |
AF332863.1 | 4 | 798 | AAL99586.1 | |
AK010765.1 | 4 | 796 | BAB27167.1 | |
AK136899.1 | 4 | 795 | BAE23161.1 | |
AK160316.1 | 4 | 797 | ||
AK162658.1 | 4 | 792 | BAE37009.1 | |
BC037239.1 | 4 | 803 | AAH37239.1 | |
BC058518.1 | 4 | 788 | AAH58518.1 | |
BC138201.1 | 4 | 883 | ||
BC145986.1 | 4 | 883 |
