Assay Details
Target Gene Details
Entrez Gene ID: | 244349 |
Gene Name: | K(lysine) acetyltransferase 6A |
Gene Aliases: |
1500036M03, 9930021N24Rik, MOZ, Myst3, Zfp220 |
Location: |
Chr.8:22859516-22943262 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kat6a | NM_001081149.1 | 3 | 636 | NP_001074618.1 |
| AK028058.1 | 2 | 555 | BAC25728.1 | |
| AK036885.1 | 3 | 631 | BAC29621.1 | |
| AK039615.1 | 3 | 634 | BAC30401.1 | |
| AK054322.1 | 3 | 630 | BAC35729.1 | |
| AK082463.1 | 3 | 637 | ||
| AK132474.1 | 2 | 544 | BAE21185.1 | |
| AK143408.1 | 3 | 627 | BAE25366.1 |
More Information
Set Membership: |
|

Genomic Map