Assay Details
Target Gene Details
Entrez Gene ID: | 74356 |
Gene Name: | RIKEN cDNA 4931428F04 gene |
Gene Aliases: |
AI426165, Kiaa0895l |
Location: |
Chr.8:105280409-105289817 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931428F04Rik | NM_001166394.1 | 7 | 1713 | NP_001159866.1 |
| NM_028888.2 | 8 | 2110 | NP_083164.1 | |
| AK016481.1 | BAB30261.1 | |||
| AK136929.1 | 7 | 1875 | BAE23173.1 | |
| BC043046.1 | 7 | 1870 | AAH43046.1 | |
| BC059045.1 | 7 | 1648 | AAH59045.1 |
Target Gene Details
Entrez Gene ID: | 78688 |
Gene Name: | nucleolar protein 3 (apoptosis repressor with CARD domain) |
Gene Aliases: |
ARC, B430311C09Rik, MYC, NOP, Nop30 |
Location: |
Chr.8:105275937-105281939 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol3 | NM_030152.4 | 4 | 2782 | NP_084428.1 |
| AK053846.1 | 4 | 2783 | BAC35554.1 |

Genomic Map