Assay Details
Target Gene Details
Entrez Gene ID: | 16336 |
Gene Name: | insulin-like 3 |
Gene Aliases: |
Rlf, Rlnl |
Location: |
Chr.8:71689252-71690577 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Insl3 | NM_013564.7 | 2 | 523 | NP_038592.3 |
AK132857.1 | 2 | 524 | BAE21395.1 | |
BC049540.1 | 2 | 523 | AAH49540.1 | |
S82815.1 | 2 | 522 | AAB39365.1 |
Target Gene Details
Entrez Gene ID: | 16453 |
Gene Name: | Janus kinase 3 |
Gene Aliases: |
fae, wil |
Location: |
Chr.8:71676383-71690577 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Jak3 | NM_010589.6 | 25 | 3870 | NP_034719.2 |
AK038268.1 | 24 | 3681 | BAC29956.1 | |
AK088365.1 | 25 | 3719 | BAC40305.1 | |
AW456897.1 | 1 | 81 | ||
BC105577.1 | 24 | 3673 | ||
L32955.1 | 24 | 3669 | AAA21565.1 |
