Assay Details
Target Gene Details
Entrez Gene ID: | 70984 |
Gene Name: | RIKEN cDNA 4931406C07 gene |
Gene Aliases: |
- |
Location: |
Chr.9:15283337-15306448 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4931406C07Rik | NM_001199484.1 | 9 | 1653 | NP_001186413.1 |
NM_001199485.1 | 9 | 1549 | NP_001186414.1 | |
NM_133732.3 | 8 | 1512 | NP_598493.1 | |
AK016432.1 | 8 | 1411 | ||
AK088123.1 | 9 | 1513 | BAC40160.1 | |
AK133192.1 | 9 | 1654 | ||
AK147073.1 | 9 | 1523 | BAE27654.1 | |
BC011540.1 | 8 | 1415 | AAH11540.1 | |
BC016078.1 | 8 | 1461 | AAH16078.1 |
