Assay Details
Target Gene Details
Entrez Gene ID: | 14065 |
Gene Name: | coagulation factor II (thrombin) receptor-like 3 |
Gene Aliases: |
PAR4 |
Location: |
Chr.8:72761186-72763898 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F2rl3 | NM_007975.3 | 2 | 399 | NP_032001.2 |
| AF080215.1 | 2 | 376 | AAC28861.1 | |
| AK036427.1 | 2 | 400 | BAC29423.1 | |
| AK153720.1 | 2 | 393 | BAE32155.1 | |
| AK156729.1 | 2 | 400 | BAE33823.1 | |
| BC119461.2 | 1 | 639 |
More Information
Set Membership: |
|

Genomic Map