Assay Details
Target Gene Details
Entrez Gene ID: | 382064 |
Gene Name: | predicted gene 1110 |
Gene Aliases: |
- |
Location: |
Chr.9:26879567-26923184 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm1110 | NM_001281475.1 | 21 | 2049 | NP_001268404.1 |
| AK136683.1 | 12 | 1475 | ||
| DQ664197.1 | 21 | 2058 |
More Information
Set Membership: |
|

Genomic Map