Assay Details
Target Gene Details
Entrez Gene ID: | 260408 |
Gene Name: | protease, serine 45 |
Gene Aliases: |
Tespl, Tessp5 |
Location: |
Chr.9:110834588-110841310 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss45 | NM_153172.1 | 4 | 509 | NP_694812.1 |
| AF392062.1 | 4 | 509 | AAM97837.1 | |
| AK077121.1 | 4 | 507 | BAC36624.1 | |
| BC107230.1 | 3 | 482 | AAI07231.1 | |
| BC107231.1 | 3 | 482 | AAI07232.1 |
More Information
Set Membership: |
|

Genomic Map