Assay Details
Target Gene Details
Entrez Gene ID: | 12049 |
Gene Name: | Bcl2-like 10 |
Gene Aliases: |
AA420380, AU023065, Boo, C85687, Diva |
Location: |
Chr.9:75347758-75351640 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcl2l10 | NM_013479.2 | 1 | 98 | NP_038507.1 |
AF067660.1 | 1 | 70 | AAC83150.1 | |
AF102501.1 | 1 | 69 | AAD08703.1 | |
AK136172.1 | 1 | 99 | BAE22856.1 | |
AK145385.1 | BAE26403.1 | |||
AK162127.1 | BAE36742.1 | |||
BC052690.1 | 1 | 97 | AAH52690.1 |
