Assay Details
Target Gene Details
Entrez Gene ID: | 69543 |
Gene Name: | calpain, small subunit 2 |
Gene Aliases: |
2310005G05Rik, 30K-2 |
Location: |
Chr.8:92901395-92902409 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Capns2 | NM_027112.1 | 1 | 176 | NP_081388.1 |
| AK009171.1 | 1 | 165 | BAB26120.1 | |
| BB571460.1 | 1 | 177 |
Target Gene Details
Entrez Gene ID: | 270084 |
Gene Name: | lysophosphatidylcholine acyltransferase 2 |
Gene Aliases: |
A330042H22, Aytl1, Aytl1a, lpafat1 |
Location: |
Chr.8:92855330-92919279 on Build GRCm38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lpcat2 | NM_173014.1 | NP_766602.1 | ||
| AB244716.1 | ||||
| AK039431.1 | BAC30345.1 | |||
| BC144806.1 | ||||
| BC150666.1 |
More Information
Set Membership: |
|

Genomic Map