Assay Details
Target Gene Details
Entrez Gene ID: | 78688 |
Gene Name: | nucleolar protein 3 (apoptosis repressor with CARD domain) |
Gene Aliases: |
ARC, B430311C09Rik, MYC, NOP, Nop30 |
Location: |
Chr.8:105275937-105281939 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol3 | NM_030152.4 | 3 | 589 | NP_084428.1 |
| AK021023.1 | 3 | 535 | BAB32281.1 | |
| AK053846.1 | 3 | 590 | BAC35554.1 | |
| AK079536.1 | 3 | 621 | BAC37674.1 | |
| AY459322.1 | 3 | 597 | AAR22550.1 | |
| BC027290.1 | 3 | 616 | AAH27290.1 |

Genomic Map