Assay Details
Target Gene Details
Entrez Gene ID: | 215418 |
Gene Name: | cysteine-serine-rich nuclear protein 1 |
Gene Aliases: |
4931429D10Rik, Axud1, CSRNP-1, URAX1, taip-3 |
Location: |
Chr.9:119971163-119985134 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Csrnp1 | NM_153287.3 | 4 | 577 | NP_695019.1 |
AB091687.1 | 3 | 549 | BAC16314.1 | |
AK029893.1 | 4 | 844 | BAC26661.1 | |
AK140213.1 | 4 | 590 | BAE24284.1 | |
AK171658.1 | 3 | 642 | ||
BC050066.1 | 4 | 577 | AAH50066.1 |
